Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ StemXVivo Chondrogenic Base Media
SDP

Catalog No. CCM005 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM005 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CCM005 Supplier R&D Systems™ Supplier No. CCM005

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Base media for the differentiation of MSCs into chondrocytes

Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into chondrocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.

  • Offers flexibility to evaluate novel cytokine and growth factor combinations to induce chondrogenesis
  • Has been developed and optimized using MSCs
  • Can be used with StemXVivo Human/Mouse or Rat Chondrogenic Supplements to reduce variation during chondrogenesis

Specifications

Product Type Stem Cell Expansion Media
Product Line R&D Systems
Quantity 50 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.