Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ StemXVivo Human/Mouse Chondrogenic Supplement, 0.5 mL
SDP

Catalog No. CCM006 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM006 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CCM006 Supplier R&D Systems™ Supplier No. CCM006

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Media supplement to induce the differentiation of MSCs into chondrocytes

Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into chondrocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.

  • Contains high quality differentiation factors to drive reproducible and efficient MSC chondrogenesis
  • Is defined to reduce unwanted experimental variability.
  • Has been developed and optimized using human and mouse MSCs

Specifications

Product Line R&D Systems
Quantity 0.5 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.