Learn More
Description
Pluripotent stem cells (PSCs) are functionally defined by their capacity for self-renewal and their ability to differentiate into cells from all three germ layers. Mouse PSCs are characterized by their marker expression, including positive markers Oct-3/4, SSEA-1, Alkaline Phosphatase, and negative marker, SSEA-4. Optimal expansion and maintenance of mouse PSCs in culture is facilitated by using specialized stem cell base media supplemented with mouse leukemia inhibitory factor (LIF).
Benefits:
- Yields highly homogenous NPC population
- Kit-derived NPCs can be differentiated into astrocytes, oligodendrocytes, and multiple neuron subtypes
Storage: Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Alternate Names: Pluripotent Stem Cells
Specifications
Specifications
| Product Type | Stem Cell Differentiation Media |
| Sterility | Sterile |
| Product Line | R&D Systems |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Source | Embryonic Stem cells |
| Content And Storage | NPC Differentiation Base Media Supplement (100X), NPC Differentiation Cocktail (1000X), Anti-Human SOX1 Antibody. Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
