Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ StemXVivo Serum-Free MSC Freezing Media
SDP

Catalog No. CCM016 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CCM016 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CCM016 Supplier R&D Systems™ Supplier No. CCM016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Freezing media for human, mouse or rat MSC cryopreservation

Mesenchymal stem/stromal cells (MSCs) are a rare population of bone marrow-derived cells. Given their rarity, MSCs are most often expanded in vitro under specific culture conditions.

  • Is defined to support reproducible MSC cryopreservation
  • Is compatible with defined, serum-free MSC culture conditions
  • Has been developed and optimized for use with human, mouse, and rat MSCs

Specifications

Product Type Freezing Media, Serum-Free
Product Line R&D Systems
Quantity 50 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.