Description
Tumorsphere assays, also called sphere-formation assays, are in vitro assays that assess stem cell characteristics of tumor cells. There are a number of variables that contribute to variation in data obtained from tumorsphere assays including the purity of the starting cell population, the type and quantity of media and supplements, and the potential for tumorspheres to aggregate.
- Shown to support sphere formation of seven tumor cell lines
- Formulated and optimized for efficient tumorsphere formation
- Lot-to-lot consistency reduces experimental variability
Specifications
Specifications
| Product Type | Stem Cell Media |
| Product Line | R&D Systems |
| Quantity | 100 mL |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |