Learn More
Sonics & Materials Inc STEPPER TIPS 1/8 (3MM)
Supplier: Sonics & Materials Inc 6300422
Stepped Microtip, 3mm, Optional Accessories For VC 750, VCX 500 and VCX 750, Compatible with most 20kHz Ultrasonic Processors