Learn More
Instech STER LOCK SOLN HEP 500USP 10ML
SDP
Supplier:  Instech USPHGS50010VBP5
Encompass_Preferred
Pharmaceutical-grade catheter lock solution prepared with heparin sodium following the USP <797> standard for compounding sterile preparations. The solution contains heparin (500 USP units/ml) in 50% glycerol, five 10 ml vials, blue, PinPort™ access.
Catalog No. NC2089607
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.