Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI STERI-BUFFER™ Phosphate Buffer
GREENER_CHOICE

Catalog No. p-7241274 Shop All VAI Products
Change view
Click to view available options
Quantity:
90 mL (3 oz.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-546-613 90 mL (3 oz.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-546-613 Supplier VAI Supplier No. SB10090
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Certificate of Analysis and Certificate of Sterility

STERI-BUFFER™ Phosphate Buffer for disinfectant preparation and dilution.

  • Buffered to a pH of 7.2 ± 0.2
  • Made with USP water for injection filtered at 0.2 μm
  • Filled in easy open-and-close wide-mouth bottles that are scaled on four sides for easy measurement
  • Terminally sterilized through a validated cycle at a SAL level of 10-6
  • Sealed with a no-tamper strip that is broken once the bottle is opened
  • Filled in a ISO 5 cleanroom
  • Validated for sterility

Order Info

Shelf life of two years. Not for parenteral administration.

Compliance

Lot sterility tested per current USP compendium.

Specifications

Chemical Name or Material Phosphate Buffer
For Use With (Application) For disinfectant preparation and dilution
pH 7.2 ± 0.2
Quantity 90 mL (3 oz.)
Sterility Sterile
Includes Certificate of Analysis, certificate of Sterility
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.