Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2 Ethylene Oxide Monitor with Acrylic Stand (Includes Remote Sensor Mount)

Catalog No. 17987115
Change view
Click to view available options
Components Measured:
Ethylene Oxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-115 Ethylene Oxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-115 Supplier ChemDAQ™ Supplier No. ST2EMONA
Add to Cart
Add to Cart
Request a Quote

Steri Trac™ 2 Ethylene Oxide Monitor with Acrylic Stand (Includes Remote Sensor Mount) provides continuous, real-time EtO detection with visual and audible alarms. Includes an acrylic stand for flexible placement and a remote sensor mount to ensure accurate monitoring at the gas source.

The Steri-Trac™ 2 Ethylene Oxide (EtO) Monitor is a continuous, fixed gas detection system designed to provide real-time monitoring of ethylene oxide vapor in sterilization and production environments. This configuration includes a clear acrylic stand for easy, portable placement on desks, carts, or workstations and a remote sensor mount for locating the sensor at the point of highest risks such as near sterilizer doors or EtO storage areas, while keeping the monitor visible and accessible. Engineered for reliability and precision, the Steri-Trac™ 2 helps protect personnel, meet OSHA and ACGIH exposure limits, and support compliance with safety protocols in healthcare, life sciences, and industrial sterilization settings.

Key Features

  • Real-Time EtO Detection Continuously monitors ambient air for the presence of ethylene oxide, providing instantaneous readings and time-weighted average (TWA) exposure data. Helps reduce the risk of overexposure and support safe re-entry after sterilization cycles.
  • Acrylic Stand for Versatile Placement The included acrylic stand provides a lightweight, non-conductive base for convenient placement on carts, tables, or workstations.
  • Remote Sensor Mount The remote sensor can be positioned independently of the display unit, allowing for accurate sampling at potential emission sources while keeping the monitor within safe, accessible areas.
  • Sensor Exchange Program (SXP™) Pre-calibrated, swappable sensor modules enable quick replacement without tools or calibration gas. Minimize downtime and ensure continuous accuracy.
  • Built-in Alarms and Indicators Multi-color LED status light and integrated buzzer provide immediate alerts for elevated EtO levels. Alarms are user-configurable to suit your facility's safety thresholds and policies.
  • Facility Integration Easily connects to building management systems (BMS) or programmable logic controllers (PLCs) via 4–20 mA, Modbus, or dry contact relays. Compatible with ChemDAQ's DAQ™ System for centralized monitoring and data logging.

Ethylene oxide is a widely used sterilant, but also a known health hazard. The Steri-Trac™ 2 EtO Monitor with Acrylic Stand provides peace of mind by continuously tracking EtO levels in critical workspaces. With real-time display, intuitive alarm features, and ChemDAQ's trusted sensor technology, this system is an essential part of any facility's safety and compliance program. Its portable configuration with an acrylic stand and remote sensor mount makes it especially useful for teams seeking flexibility, precision, and quick deployment, without compromising on performance or integration capabilities.

Specifications

Components Measured Ethylene Oxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.