Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Ethylene Oxide Monitor with Acrylic Stand (Includes Remote Sensor Mount)

Catalog No. 17987122
Change view
Click to view available options
Components Measured:
Ethylene Oxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-122 Ethylene Oxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-122 Supplier ChemDAQ™ Supplier No. ST2PEMONA
Add to Cart
Add to Cart

Steri-Trac™ 2+ Ethylene Oxide Monitor with acrylic stand offers continuous, real-time EtO monitoring. Includes remote sensor mount for accurate detection at the source. Ideal for SPDs, labs, and sterilization areas needing flexible, portable safety monitoring.

The Steri-Trac™ 2+ Ethylene Oxide (EtO) Monitor is a fixed, real-time gas detection system designed to continuously monitor airborne levels of ethylene oxide in sterilization and industrial environments. This configuration includes a durable acrylic stand for convenient desktop or cart placement, and a remote sensor mount to position the sensor directly in high-risk zones. Built for performance, safety, and integration, the Steri-Trac™ 2+ delivers live exposure data, customizable alarms, and communication capabilities to support compliance with occupational safety standards.

Key Features

  • Real-Time EtO Monitoring Continuously measures ambient ethylene oxide levels with instantaneous, TWA, and STEL readings helping protect staff and support OSHA, NIOSH, and ACGIH exposure compliance.
  • Portable Acrylic Stand The included clear acrylic stand allows easy placement on desks, workstations, or mobile carts.
  • Remote Sensor Mount The sensor can be installed separately from the monitor, allowing detection at the point of highest potential EtO exposure while keeping the monitor accessible for staff.
  • SXP™ Sensor Exchange Program Utilizes ChemDAQ's SXP™ pre-calibrated sensors for quick, tool-free replacements. No calibration gas or downtime required.
  • Customizable Alarms Integrated visual and audible alarms notify users when EtO levels exceed defined thresholds. Alarm points are easily configurable to match safety protocols.
  • Integration-Ready Supports Modbus, 4–20 mA, and dry contact relays for seamless connection to facility management systems, abatement controls, or ChemDAQ's DAQ™ data platform.

Ethylene oxide is an essential sterilant, but also a hazardous chemical that demands close monitoring. The Steri-Trac™ 2+ with Acrylic Stand and Remote Sensor Mount offers unmatched flexibility, real-time protection, and ease of integration. Whether deployed in a hospital SPD, manufacturing floor, or sterilization area, it helps ensure a safe working environment and continuous regulatory compliance.

Specifications

Components Measured Ethylene Oxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.