Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2 Ethylene Oxide Monitor with Steel Swivel Wall Mount (Includes Remote Sensor Mount)

Catalog No. 17987116
Change view
Click to view available options
Components Measured:
Ethylene Oxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-116 Ethylene Oxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-116 Supplier ChemDAQ™ Supplier No. ST2EMONW
Add to Cart
Add to Cart
Request a Quote

Steri Trac™ 2 Ethylene Oxide Monitor with Steel Swivel Wall Mount (Includes Remote Sensor Mount) delivers continuous, real-time EtO detection with audible/visual alarms. The robust steel swivel mount ensures secure, adjustable placement, and the remote sensor mount allows flexible placement at the source.

Steri-Trac™ 2 Ethylene Oxide Monitor with Steel Swivel Wall Mount (Includes Remote Sensor Mount) The Steri-Trac™ 2 Ethylene Oxide (EtO) Monitor is a fixed, real-time gas monitoring system designed to detect and display airborne ethylene oxide concentrations in workplace environments. This version includes a durable steel swivel wall mount, offering secure and adjustable installation, and a remote sensor mount that allows the sensing element to be placed at the source of potential exposure for more accurate readings. Ideal for healthcare, pharmaceutical, and industrial sterilization settings, the Steri-Trac™ 2 continuously monitors EtO levels to help facilities maintain a safe working environment and ensure regulatory compliance.

Key Features:

  • Real-Time Monitoring Continuously displays instantaneous ethylene oxide (EtO) concentrations in parts per million (ppm) for rapid detection of unsafe levels.
  • Remote Sensor Mount Included Enables the sensor to be positioned near potential gas sources (e.g., sterilizers, aeration rooms) while keeping the main display unit mounted safely and conveniently on the wall.
  • Steel Swivel Wall Mount Heavy-duty mount allows the monitor to be securely installed and easily adjusted for optimal visibility and access in high-traffic or restricted spaces.
  • Visual & Audible Alarms Tri-color LED indicators (green/yellow/red) and built-in audible alerts provide immediate notification of gas levels exceeding user-defined thresholds, helping prevent overexposure incidents.
  • Sensor Exchange Program (SXP™) Compatible Supports ChemDAQ's Sensor Exchange Program, enabling easy replacement of pre-calibrated sensor modules to minimize downtime and maintenance.
  • Time-Weighted Average (TWA) Readings Calculates and displays 8-hour and 15-minute TWAs.
  • Data Integration Can be integrated with the ChemDAQ™ Data Acquisition System (DAQ™) or connected to facility systems via Modbus, 4–20 mA analog outputs, or dry contact relays for alarm/abatement activation and logging.
  • The Steri-Trac™ 2 Ethylene Oxide Monitor with Steel Swivel Wall Mount is a reliable, safety-first solution for facilities that require constant vigilance against ethylene oxide exposure. Its rugged design, real-time accuracy, and flexible mounting make it a trusted choice for compliance-driven, high-risk environments.

Specifications

Components Measured Ethylene Oxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.