Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Ethylene Oxide Monitor with Steel Swivel Wall Mount (Includes Remote Sensor Mount)

Catalog No. 17987123
Change view
Click to view available options
Components Measured:
Ethylene Oxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-123 Ethylene Oxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-123 Supplier ChemDAQ™ Supplier No. ST2PEMONW
Add to Cart
Add to Cart

Steri-Trac™ 2+ Ethylene Oxide Monitor with steel swivel wall mount provides continuous, real-time EtO gas monitoring. Includes remote sensor mount for flexible placement and accurate detection at the source. Ideal for sterilization and industrial safety.

The Steri-Trac™ 2+ Ethylene Oxide (EtO) Monitor is a fixed, continuous gas detection system designed for real-time monitoring of ethylene oxide levels in sterilization rooms, industrial facilities, and other critical environments. This model includes a robust steel swivel wall mount for secure, adjustable installation on walls or equipment, and a remote sensor mount to position the sensor near potential exposure points while keeping the monitor accessible. Engineered for reliability, safety, and seamless integration, the Steri-Trac™ 2+ delivers accurate exposure data and alerts to help protect workers and maintain regulatory compliance.

Key Features

  • Real-Time Ethylene Oxide Monitoring Continuously measures ambient EtO gas concentrations, displaying instantaneous, time-weighted average (TWA), and short-term exposure limit (STEL) values to ensure safe workplace air quality.
  • Steel Swivel Wall Mount Heavy-duty swivel mount offers flexible installation options on walls or machinery, allowing precise positioning and easy viewing while saving floor space.
  • Remote Sensor Mount Place the sensor directly where EtO exposure risk is highest near sterilizers, vents, or storage areas while the monitor remains in a safe, convenient location.
  • SXP™ Sensor Exchange Program Utilizes ChemDAQ's pre-calibrated, tool-free replaceable SXP™ sensors, minimizing downtime and simplifying maintenance.
  • Configurable Visual and Audible Alarms Bright LED indicators and loud alarms provide immediate notification of unsafe EtO levels. Alarm thresholds can be customized to fit site-specific safety protocols.
  • Facility Integration Compatible Supports industrial communication standards including Modbus RTU, 4–20 mA analog output, and dry contact relays, enabling connection to building management systems (BMS), programmable logic controllers (PLCs), and abatement equipment.

Protecting personnel from ethylene oxide exposure requires reliable, real-time monitoring in critical areas. The Steri-Trac™ 2+ with its durable steel swivel wall mount and remote sensor capability offers flexibility, precision, and compliance-ready features tailored for hospital sterilization areas, pharmaceutical production, and other industrial environments. Its advanced sensor technology and integration options make it a trusted solution to ensure a safer workplace.

Specifications

Components Measured Ethylene Oxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.