Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Hydrogen Peroxide Monitor with Acrylic Stand

Catalog No. 17987124
Change view
Click to view available options
Components Measured:
Hydrogen Peroxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-124 Hydrogen Peroxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-124 Supplier ChemDAQ™ Supplier No. ST2PHMONA
Add to Cart
Add to Cart
Request a Quote

Steri-Trac™ 2+ Hydrogen Peroxide Monitor offers real-time H2O2 detection with visual/audible alarms. Includes an acrylic stand for easy placement. Ideal for monitoring sterilization areas and ensuring workplace safety.

The Steri-Trac™ 2+ Hydrogen Peroxide (H2O2) Monitor is a high-precision, fixed gas detection system that provides continuous, real-time monitoring of hydrogen peroxide vapor levels. Ideal for use in environments where H2O2 is used for disinfection, sterilization, or decontamination, this configuration includes a durable acrylic stand for easy placement on desks, carts, or workstations, and a remote sensor mount for optimized monitoring at the point of potential exposure. Designed for use in healthcare, pharmaceutical, and laboratory settings, the Steri-Trac™ 2+ offers the advanced performance and integration capabilities required to protect staff and maintain safety compliance.

Key Features

  • Continuous Hydrogen Peroxide Monitoring Provides accurate, real-time measurement of airborne H2O2 levels. Displays instantaneous, TWA (time-weighted average), and STEL (short-term exposure limit) data to support OSHA and ACGIH compliance.
  • Portable Acrylic Stand The included acrylic stand allows for flexible and stable placement on counters, workstations, or mobile carts.
  • Remote Sensor Mount Place the sensor closer to the point of emission (e.g., sterilizers, decontamination chambers, or room vents) for accurate detection, while the monitor remains easily accessible.
  • SXP™ Sensor Exchange Program ChemDAQ's SXP™ system enables quick, pre-calibrated sensor replacements with no downtime, calibration gas, or technical service required.
  • Visual and Audible Alarms Integrated, configurable alarms immediately alert staff to unsafe H2O2 levels through bright LED lights and a loud buzzer, ensuring rapid response.
  • Seamless Facility Integration Supports 4–20 mA, Modbus, and dry contact relays for integration with building management systems (BMS), alarms, and automation systems.

Hydrogen peroxide is a powerful sterilant, but overexposure can pose serious health risks. The Steri-Trac™ 2+ H2O2 Monitor delivers high-performance monitoring with the flexibility of desktop deployment and the precision of remote sensor placement. Whether used in sterilization rooms, cleanrooms, or research facilities, this system provides a reliable safeguard against airborne H2O2, keeping your personnel safe and your operations compliant.

Specifications

Components Measured Hydrogen Peroxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.