Learn More
ChemDAQ™ Steri-Trac™ 2 Hydrogen Peroxide Monitor with Steel Swivel Wall Mount
Description
- Continuously displays real-time measurements of vapor concentrations in parts per million (ppm)
- Displays 15-min and 8-hr Time Weighted Average (TWA) exposure readings
- Customizable audible and visual alarms, including impending alerts
- Timely and consistent shipment of calibrated sensors through ChemDAQ’s exclusive Sensor Exchange Program (SXP™)
Specifications
Specifications
| Components Measured | Hydrogen Peroxide |
| For Use With (Equipment) | ChemDAQ Gas Sensor |
| Type | Fixed Gas Detection Monitor |
Purchase of this product requires an annual service contract for sensor calibration and exchange, which must be arranged prior to shipment. Customers in the food and beverage production and processing industry need to contact ChemDAQ directly to purchase this product. Contact your Fisher Scientific representative to confirm your needs and to request a quote.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.