Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2 Intrinsically Safe Oxygen Package - includes Monitor, Barrier Unit, and Remote Sensor Mount

Catalog No. 17987121
Change view
Click to view available options
Components Measured:
Oxygen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-121 Oxygen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-121 Supplier ChemDAQ™ Supplier No. ST2ISO2PKG
Add to Cart
Add to Cart
Request a Quote

Steri-Trac™ 2 Intrinsically Safe Oxygen Package provides continuous O2 monitoring in hazardous areas. Includes monitor, intrinsically safe barrier unit, and remote sensor mount for safe, accurate detection. Ideal for classified environments and oxygen-sensitive operations.

The Steri-Trac™ 2 Intrinsically Safe (IS) Oxygen Monitoring Package is a fixed, real-time gas detection system engineered for continuous monitoring of oxygen (O2) levels in hazardous or classified environments, such as sterilization rooms, cleanrooms, or chemical storage areas. Designed with an Intrinsically Safe Barrier Unit and a Remote Sensor Mount, this package meets the strict safety requirements of Class I, Division 1 or Zone 0 locations where flammable gases may be present. The system helps protect personnel from both oxygen-enriched and oxygen-deficient conditions by providing constant monitoring, customizable alarms, and seamless integration with safety systems.

Key Features

  • Continuous Oxygen Monitoring Provides accurate, real-time measurement of ambient oxygen levels, helping ensure safe breathing air and preventing combustion risks associated with high oxygen concentrations.
  • Intrinsically Safe Barrier Unit Included to safely operate in explosive or flammable atmospheres, the IS Barrier Unit restricts electrical energy to the sensor, making the system compliant with hazardous environment standards.
  • Remote Sensor Mount Place the oxygen sensor directly where air composition is most critical, such as near gas lines, tanks, or sterilizers.
  • SXP™ Sensor Technology Uses ChemDAQ's Sensor Exchange Program (SXP™) for fast, tool-free sensor replacements. Pre-calibrated modules reduce maintenance time and eliminate downtime.
  • Visual & Audible Alarms Bright LED indicators and a loud buzzer alert staff to unsafe oxygen levels. Alarm thresholds are fully configurable to match site-specific protocols.
  • Facility Integration Ready Connects easily with building management systems (BMS), programmable logic controllers (PLCs), and safety interlocks via 4–20 mA, Modbus, and dry contact relays.

Maintaining safe oxygen levels is critical in hazardous environments, where both oxygen depletion and enrichment pose serious safety risks. The Steri-Trac™ 2 IS Oxygen Package delivers accurate, real-time monitoring and robust alarm functionality in a configuration that meets the highest safety standards. Its intrinsically safe design, flexible sensor placement, and industry-leading ChemDAQ sensor technology make this package the ideal choice for facilities handling gases, sterilants, or working in confined spaces where atmosphere control is essential.

Specifications

Components Measured Oxygen
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.