Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Intrinsically Safe Package - includes Monitor, Barrier Unit, and Remote Sensor Mount

Catalog No. 17987126
Change view
Click to view available options
Components Measured:
Ethylene Oxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-126 Ethylene Oxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-126 Supplier ChemDAQ™ Supplier No. ST2PISEPKG
Add to Cart
Add to Cart
Request a Quote

Steri-Trac™ 2+ Intrinsically Safe Package provides real-time gas monitoring in hazardous areas. Includes monitor, intrinsically safe barrier unit, and remote sensor mount for safe detection in classified environments. Ideal for explosion-prone industrial or sterilization settings.

The Steri-Trac™ 2+ Intrinsically Safe Package is a complete, fixed gas monitoring system designed for real-time detection of hazardous gases in classified and potentially explosive environments. Built to meet rigorous industrial and safety standards, this package includes the Steri-Trac 2+ gas monitor, an Intrinsically Safe (IS) barrier unit, and a remote sensor mount, enabling flexible and safe deployment in critical zones. Whether you're monitoring sterilant gases like Ethylene Oxide (EtO), Peracetic Acid (PAA), Hydrogen Peroxide (H2O2), or flammable gases near their Lower Explosive Limits (LEL), this intrinsically safe package is ideal for industrial, laboratory, and healthcare environments that demand the highest levels of protection and performance.

Key Features

  • Intrinsically Safe Design: The package includes an IS barrier that safely limits electrical energy in hazardous areas, preventing ignition of flammable atmospheres. Certified for use in Class I, Division 1 or 2 environments, it enables safe operation in facilities where explosive gases may be present.
  • Continuous, Real-Time Monitoring: The Steri-Trac 2+ monitor continuously tracks gas levels and displays instantaneous concentrations, as well as time-weighted averages (TWA) and short-term exposure limits (STEL) when applicable. This provides safety personnel with comprehensive exposure information.
  • Remote Sensor Mount: To optimize sensor placement, the remote sensor mount allows the gas sensor to be positioned at the highest risk, such as near sterilizers, fume hoods, tanks or valves.
  • Sensor Exchange Program (SXP) Compatible: The monitor is designed to work with ChemDAQ's Sensor Exchange Program (SXP), which provides pre-calibrated, hot-swappable sensors. This eliminates the need for on-site calibration, reduces downtime, and ensures continuous accuracy with minimal maintenance.
  • Multi-Level Alarm System: Equipped with a multi-color LED indicator and audible alarm, the monitor alerts users when gas levels exceed defined thresholds. Alarms are fully configurable, allowing you to tailor the system to your facility's safety protocols and compliance requirements.
  • Seamless System Integration: The Steri-Trac™ 2+ supports integration with building management systems, safety interlocks, or facility alarms using 4-20 mA output, Modbus, and dry contact relays. This enables automation of response measures like ventilation, lockdowns, or evacuation procedures.

For facilities that operate in hazardous environments where flammable gases or sterilants are used, safety and compliance are non-negotiable. The Steri-Trac™ 2+ Intrinsically Safe Package delivers advanced sensing, reliable alarm notification, and trusted protection backed by ChemDAQ's expertise in industrial gas monitoring. This package combines sensor flexibility, explosion-proof assurance, and integration readiness, making it the ideal choice for EHS professionals, safety officers, and facility managers who need dependable gas monitoring in high-risk areas.

Specifications

Components Measured Ethylene Oxide
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.