Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Intrinsically Safe Package for Lower Explosive Limit - includes Monitor, Barrier Unit, and Remote Sensor Mount

Catalog No. 17987127
Change view
Click to view available options
Components Measured:
LEL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-127 LEL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-127 Supplier ChemDAQ™ Supplier No. ST2PISLPKG
Add to Cart
Add to Cart
Request a Quote

Steri-Trac™ 2+ Intrinsically Safe Package for Lower Explosive Limit (LEL) includes a monitor, barrier unit, and remote sensor mount. Provides real-time LEL monitoring in hazardous areas, ensuring safety and compliance in explosive or flammable environments.

The Steri-Trac™ 2+ Intrinsically Safe Package for LEL monitoring is a specialized gas detection solution designed for real-time detection of combustible gases at concentrations approaching the Lower Explosive Limit (LEL). This complete safety package includes a Steri-Trac™ 2+ Monitor, Intrinsically Safe (IS) Barrier Unit, and a Remote Sensor Mount, making it ideal for use in Class I, Division 1 or 2 hazardous locations where the potential for flammable gas accumulation exists. Designed to protect personnel and facilities from explosion hazards, this system is widely used in chemical processing, manufacturing, sterilization environments using flammable gases, and other classified industrial areas.

Key Features

  • Real-Time LEL Detection: Continuously monitors the air for the presence of flammable gases and vapors and displays their concentration as a percentage of the LEL. The system provides rapid alerts if gas levels approach dangerous thresholds, enabling fast evacuation or mitigation response.
  • Intrinsically Safe Design: The included IS Barrier Unit ensures that electrical energy entering the hazardous area is limited to safe levels, preventing sparks or heat that could ignite flammable gases. The entire system is compliant with standards for use in explosion-prone environments.
  • Remote Sensor Mount: The gas sensor can be mounted remotely close to the area with the highest risk of gas buildup, while keeping the monitor and display in a safe, accessible area. Ideal for monitoring around sterilizers, equipment vents, gas storage, and confined spaces.
  • Sensor Exchange Program (SXP™): Designed to work with ChemDAQ's SXP™ system, the sensor can be hot swapped with a pre-calibrated replacement, significantly reducing downtime and eliminating the need for on-site calibration.
  • Customizable Alarms: Bright tri-color LEDs and a high-decibel buzzer provide both visual and audible alerts. Users can configure alarms based on their facility's specific safety standards, including low-level warnings and high-level emergency thresholds.
  • System Integration Ready: The monitor supports 4–20 mA analog output, Modbus, and dry contact relays for connection to building management systems (BMS), safety interlocks, emergency shutoff valves, and ChemDAQ's Data Acquisition System (DAQ).

The Steri-Trac™ 2+ Intrinsically Safe Package for LEL is ChemDAQ's premier solution for protecting lives and property from the dangers of flammable gas accumulation. With continuous monitoring, smart integration options, and certified explosion protection, it is ideal for organizations that need serious gas safety in classified zones. From hospitals and sterile processors to chemical labs and gas storage facilities, the Steri-Trac™ 2+ ensures your team is alerted to combustible risks before they escalate, keeping your operations safe, compliant, and efficient.

Specifications

Components Measured LEL
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.