Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Peracetic Acid Monitor with Acrylic Stand

Catalog No. 17987129
Change view
Click to view available options
Components Measured:
Peracetic Acid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-129 Peracetic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-129 Supplier ChemDAQ™ Supplier No. ST2PMONA
Add to Cart
Add to Cart
Request a Quote

Steri-Trac™ 2+ Peracetic Acid Monitor provides real-time PAA detection with visual/audible alarms. Includes an acrylic stand for flexible placement. Ideal for monitoring sterilization areas and ensuring safe working conditions.

The Steri-Trac™ 2+ Peracetic Acid (PAA) Monitor with acrylic stand is a compact, reliable, and easy-to-deploy fixed gas detection system designed to provide real-time, continuous monitoring of airborne peracetic acid, a powerful disinfectant widely used in healthcare, pharmaceutical, biotech, food processing, and laboratory environments. This configuration includes a clear acrylic stand, allowing for freestanding placement on desks, carts, counters, or other flat surfaces. It's ideal for facilities needing flexible installation without committing to wall mounting, especially in workstations, sterilization prep rooms, or temporary disinfection areas.

Key Features

  • Real-Time PAA Detection The Steri-Trac™ 2+ continuously measures peracetic acid in the air, displaying concentrations in parts per million (ppm) to help teams detect and address unsafe conditions immediately. Audible and visual alarms activate when pre-set exposure thresholds are exceeded, providing instant safety alerts.
  • Freestanding Acrylic Stand This version includes a durable, transparent acrylic stand that allows for flexible and mobile installation. The stand is ideal for temporary setups or for users who need to reposition the monitor as facility needs change, without permanent mounting or hardware.
  • Time-Weighted Averages (TWAs) and Short-Term Exposure Limits (STELs) To help ensure compliance with OSHA, ACGIH, and NIOSH guidelines, the monitor automatically calculates and displays:
    • 8-hour Time-Weighted Average (TWA)
    • 15-minute Short-Term Exposure Limit (STEL)
    • This gives staff and safety managers full visibility into both real-time and cumulative exposure levels.
  • Sensor Exchange Program (SXP™) The monitor is compatible with ChemDAQ's hot-swappable, pre-calibrated sensor modules through the Sensor Exchange Program (SXP™). This eliminates the need for on-site calibration and ensures minimal downtime for sensor replacement and maintenance.
  • Configurable Alarms The system is equipped with multi-color LED indicators and a built-in buzzer. Alarms can be fully configured based on your facility's safety policies and local regulations.
  • Easy System Integration This model is integration-ready with support for:
    • Modbus
    • 4–20 mA analog output
    • Dry contact relays Use these outputs to trigger external alarms, activate ventilation systems, or feed data into building management systems (BMS), safety interlocks, or ChemDAQ's own Data Acquisition System (DAQ™).

The Steri-Trac™ 2+ is trusted by hospitals, manufacturers, and laboratories across the globe. Its high accuracy, flexible deployment, and integration capabilities make it one of the most advanced solutions for sterilant gas detection. With the acrylic stand configuration, you get the added benefit of fast, non-permanent setup that allows your safety team to respond quickly to emerging risks or changing workflows. Whether you're adding temporary monitoring to a sterilization area or equipping a mobile disinfection station, this unit offers the performance and adaptability you need.

Specifications

Components Measured Peracetic Acid
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.