Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Peracetic Acid Monitor with Acrylic Stand

Catalog No. 17987131
Change view
Click to view available options
Components Measured:
Peracetic Acid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-131 Peracetic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-131 Supplier ChemDAQ™ Supplier No. ST2PPMONA
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Steri-Trac™ 2+ Peracetic Acid Monitor delivers real-time PAA monitoring with visual/audible alarms. Includes an acrylic stand for convenient placement. Ideal for ensuring safety in sterilization and cleanroom environments.

The Steri-Trac™ 2+ Peracetic Acid (PAA) Monitor is a fixed, real-time gas detection system designed for continuous monitoring of airborne peracetic acid, a commonly used chemical disinfectant in healthcare, pharmaceutical, and food processing environments. This version includes a transparent acrylic stand, allowing for easy, freestanding placement on desks, counters, or carts. Whether monitoring high-level disinfection areas or ensuring safe reentry after sterilant use, the Steri-Trac™ 2+ provides immediate, reliable readings to protect workers and maintain compliance with regulatory safety limits.

Key Features

  • Real-Time PAA Detection The monitor continuously measures peracetic acid concentrations in the air and displays real-time levels in parts per million (ppm). It helps prevent overexposure by alerting personnel to hazardous conditions as soon as they occur.
  • Acrylic Stand for Portable Placement Included acrylic stand allows flexible, non-permanent installation. Perfect for use on workbenches, equipment carts, or areas where wall-mounting is not practical. The stand's clear design blends into sterile or clinical environments without taking up significant space.
  • Time-Weighted Averages The Steri-Trac™ 2+ also calculates and displays 15-minute short-term exposure limits (STEL) and 8-hour time-weighted averages (TWA) to support regulatory compliance with OSHA, AAMI, and NIOSH exposure limits for peracetic acid.
  • Sensor Exchange Program (SXP™) Compatible with ChemDAQ's SXP™ system, this monitor allows quick replacement of pre-calibrated, hot-swappable sensor modules, which ensures accuracy with minimal downtime or need for on-site calibration.
  • Audible and Visual Alarms The monitor features tri-color LED status indicators and a built-in audible alarm to alert staff when gas concentrations exceed pre-set limits. Alarms can be customized to meet site-specific safety policies.
  • Data and System Integration The unit can be connected to ChemDAQ's Data Acquisition System (DAQ™) or third-party systems using Modbus, 4–20 mA analog output, or dry contact relays, making it easy to integrate into existing facility safety networks or building management systems (BMS).

ChemDAQ's Steri-Trac™ systems are trusted worldwide in industries that rely on sterilant chemicals. The Steri-Trac™ 2+ PAA Monitor with Acrylic Stand offers the perfect combination of flexibility, accuracy, and user-friendly design. It's ideal for both temporary and long-term installations, offering real-time safety data that helps teams work confidently in environments where peracetic acid is used.

Specifications

Components Measured Peracetic Acid
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.