Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Peracetic Acid Monitor with Steel Swivel Wall Mount

Catalog No. 17987130
Change view
Click to view available options
Components Measured:
Peracetic Acid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-130 Peracetic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-130 Supplier ChemDAQ™ Supplier No. ST2PMONW
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Steri-Trac™ 2+ Peracetic Acid Monitor with Steel Swivel Wall Mount provides real-time PAA detection with visual/audible alarms. Includes a durable steel swivel wall mount for secure, adjustable placement in sterilization and processing areas.

The Steri-Trac™ 2+ Peracetic Acid (PAA) Monitor is a continuous, fixed gas detection system designed to provide real-time measurement of airborne peracetic acid levels in critical work environments. Engineered for reliability, safety, and regulatory compliance, this model includes a steel swivel wall mount, offering a secure, adjustable installation ideal for permanent monitoring in sterilization, disinfection, and cleanroom areas. Whether you're operating in healthcare, pharmaceutical production, food processing, or laboratory environments, the Steri-Trac™ 2+ delivers accurate, uninterrupted detection of PAA to help you maintain a safe workplace and prevent harmful exposure.

Key Features

  • Real-Time Peracetic Acid Monitoring Continuously displays current PAA concentrations in parts per million (ppm), helping you detect exposure risks as they occur. Built-in visual and audible alarms notify staff when levels exceed user-defined safety thresholds.
  • Durable Steel Swivel Wall Mount This model includes a robust steel swivel mount that allows the monitor to be securely attached to a wall while offering flexible adjustment for improved visibility and access. Ideal for permanent installations where space optimization and equipment accessibility are critical.
  • TWA and STEL Exposure Monitoring Displays 15-minute short-term exposure limits (STELs) and 8-hour time-weighted averages (TWAs) to assist with OSHA, ACGIH, and NIOSH exposure limit compliance.
  • Hot-Swappable Sensors with SXP™ Compatible with ChemDAQ's Sensor Exchange Program (SXP™), enabling fast, no-calibration sensor replacement. Pre-calibrated sensors reduce downtime and make maintenance simple and efficient.
  • Customizable Visual & Audible Alarms Features bright multi-color LED status indicators (green/yellow/red) and a built-in buzzer. Alarms can be configured to match your facility's safety thresholds and policies.
  • Advanced Integration Capabilities Supports standard industrial protocols including Modbus, 4–20 mA analog output, and dry contact relays for seamless integration into facility safety systems, abatement controls, or building management systems (BMS).

ChemDAQ is the global leader in sterilant gas monitoring systems. With proven reliability in healthcare, pharma, and industrial settings, the Steri-Trac™ 2+ offers unmatched precision, safety, and ease of use. The steel swivel wall-mounted version is perfect for facilities that require fixed-position, professional-grade monitoring of peracetic acid, with smart integration and long-term value. Whether you're upgrading from an older system or building a new sterile processing area, this monitor provides everything you need to protect your staff, meet regulatory standards, and ensure continuous safety.

Specifications

Components Measured Peracetic Acid
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.