Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChemDAQ™ Steri-Trac™ 2+ Peracetic Acid Monitor with Steel Swivel Wall Mount

Catalog No. 17987132
Change view
Click to view available options
Components Measured:
Peracetic Acid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-987-132 Peracetic Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-987-132 Supplier ChemDAQ™ Supplier No. ST2PPMONW
Add to Cart
Add to Cart
Request a Quote

Steri-Trac™ 2+ Peracetic Acid Monitor with Steel Swivel Wall Mount offers real-time PAA detection with visual and audible alarms. The durable steel swivel mount allows secure, adjustable placement which is ideal for fixed monitoring in sterilization and processing areas.

The Steri-Trac™ 2+ Peracetic Acid (PAA) Monitor is a fixed, real-time gas detection system designed to measure airborne concentrations of peracetic acid used in sterilization and disinfection processes. This version comes with a durable steel swivel wall mount, providing a secure, adjustable installation for optimal placement in high-traffic or hard-to-reach areas. Used widely in healthcare, pharmaceutical, food processing, and cleanroom environments, this monitor helps facilities maintain a safe working environment by offering accurate, continuous readings of PAA levels and alerting staff before exposure reaches hazardous levels.

  • Real-Time Peracetic Acid Monitoring The Steri-Trac™ 2+ continuously detects and displays peracetic acid levels in parts per million (ppm). It provides immediate visual and audible alarms when preset thresholds are exceeded, helping prevent overexposure and maintain compliance with health and safety standards.
  • Steel Swivel Wall Mount This version includes a heavy-duty steel swivel mount that allows the monitor to be securely fixed to a wall while enabling flexible adjustment. The swivel feature makes it easy to reposition the display unit for visibility, access, or space constraints.
  • Remote Sensor Compatibility The system is compatible with ChemDAQ's remote sensor mount (sold separately), allowing the sensor to be installed closer to the point of exposure, such as in sterilizers, cleanrooms, or disinfection chambers.
  • Instantaneous & Time-Weighted Average (TWA) Readings In addition to real-time concentration display, the monitor calculates 8-hour and 15-minute TWAs, supporting compliance with regulatory bodies such as OSHA, ACGIH, and NIOSH.
  • Sensor Exchange Program (SXP™) The monitor supports ChemDAQ's Sensor Exchange Program, which allows for quick replacement of pre-calibrated, hot-swappable sensor modules. This minimizes downtime, eliminates the need for in-house calibration, and ensures ongoing measurement accuracy.
  • Customizable Visual and Audible Alarms With multicolor LED indicators (green/yellow/red) and a built-in buzzer, the monitor provides clear, immediate alerts for elevated PAA levels. Alarm settings are customizable based on facility thresholds and operational requirements.
  • System Integration The Steri-Trac™ 2+ can be integrated with facility safety systems using Modbus, 4–20 mA analog outputs, or dry contact relays. This enables automatic activation of ventilation, alarms, abatement systems, or other safety controls when PAA levels rise.

ChemDAQ has been a trusted name in gas monitoring systems for sterilant chemicals for over two decades. The Steri-Trac™ 2+ represents the next generation of monitoring systems, designed to meet modern facility demands for accuracy, flexibility, compliance, and ease of use. Backed by ChemDAQ's customer support and Sensor Exchange Program, it is an industry-leading solution for real-time PAA monitoring.

Specifications

Components Measured Peracetic Acid
For Use With (Equipment) ChemDAQ Gas Sensor
Type Fixed Gas Detection Monitor

Purchase of this product requires and annual service contract for sensor calibration and exchange. Contact your Fisher Scientific representative to confirm your needs and provide a quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.