Learn More
Dukal™ Sterile Alcohol Prep Pads
Description
- Saturated with 70% isopropyl alcohol
- Ideal for use as an antiseptic cleanser
- 2-ply, non-woven pads
- Not made with natural rubber latex
Specifications
Specifications
| Product Type | Alcohol Prep Pad |
| Active Ingredients | 70% Isopropyl alcohol |
| Description | Non-woven; 2-ply |
| For Use With (Application) | Antiseptic Cleanser |
| Latex Free | Yes |
| Quantity | 1000/Pk., 4 Pk./Cs. |
| Size | Medium |
| Sterility | Sterile |