Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems STERILE BULK SORT CARTRDG 10PK
SDP

Numéro de catalogue. NC2309428 missing translation for 'brandsLinkText'
Encompass

  • Provides a robust solution for bulk cell sorting
  • Individually wrapped for sterility
  • Compatible with Hana and Pala cell sorters
  • Processes cell samples up to 600 µL
  • Optimal sorting rate of up to 50 events/second
  • Handles cell concentrations from 100,000-500,000 cells/mL
  • Enrichment capabilities for higher concentrations up to 150M cells/mL
  • Achieves output purities of up to 90%
  • Suitable for research use only
  • Shipping at ambient temperature
  • Storage at room temperature (unopened product)
  • Applications include cell sorting and single cell dispensing

Numéro de catalogue. NC2309428 Fournisseur R&D Systems Code fournisseur NC101
May include imposed supplier surcharges.
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

  • Provides a robust solution for bulk cell sorting
  • Individually wrapped for sterility
  • Compatible with Hana and Pala cell sorters
  • Processes cell samples up to 600 µL
  • Optimal sorting rate of up to 50 events/second
  • Handles cell concentrations from 100,000-500,000 cells/mL
  • Enrichment capabilities for higher concentrations up to 150M cells/mL
  • Achieves output purities of up to 90%
  • Suitable for research use only
  • Shipping at ambient temperature
  • Storage at room temperature (unopened product)
  • Applications include cell sorting and single cell dispensing

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.