Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Superior Glove™ Sterile Cleanroom Cut-Resistant Knit Gloves

Catalog No. 17489627
Change view
Click to view available options
Size:
8
11
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-489-627 8
17-489-630 11
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-489-627 Supplier Superior Glove™ Supplier No. CRSSXPUQ8

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Cleanroom processed to minimize contamination from dust and particles for industry uses in pharmaceutical, electronic assembly and automotive paint line

  • Dyneema™ is lint-free and provides cut and abrasion resistance
  • Economy version. 44 grams/pr
  • Polyurethane provides secure grip in wet, oily conditions
  • Form fitting, breathable, and flexible gloves are ergonomically shaped
  • High dexterity and flexibility Washable and bleachable (under 145°C)
  • ASTM ANSI Cut Level 2

Specifications

Color White
Product Type Cut-resistant Gloves
Size 8
Sterility Sterile
For Use With (Application) Cleanroom environment
Glove Cuff Style Knit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.