Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Sterile Forceps
SDP

Catalog No. 22130489
Change view
Click to view available options
Quantity:
100/Cs.
25/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-130-489 25/Cs.
22-130-515 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-130-489 Supplier StatLab Supplier No. 900
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cost-effective pre-sterilized stainless steel forceps.

  • Rounded, grooved tip enhances ability to pick up and hold small objects
  • Reduce exposure to infectious disease with disposable instruments
  • Sturdy 4.5 in. long stainless steel
  • Individually wrapped sterile packets

Specifications

Product Type Forceps
Length (English) 4.5 in.
Length (Metric) 114.3 mm
Material Stainless Steel
Quantity 25/Cs.
Style Curved
Tip Style Grooved
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.