Learn More
Stoelting™ Sterile Gauze
SDP
Stoelting™ Sterile Gauze provides strength, absorbency and scrubbing texture
Supplier:  Stoelting™ 50968
 View more versions of this product
Catalog No. 10-000-685
Description
Description
  • Protects and cushions wounds
  • Promotes quick drainage and full utilization of entire sponge
  • 100% cotton
  • Peel-open package ensures sterile integrity and aseptic preparation
  • Edges are folded inward to eliminate linting and fraying
  • Unique rayon and polyester nonwoven fabric
Specifications
Specifications
Neuroscience
2 x 2 in.
Synthetic gauze
Case of 1500
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.