Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Sterile PET Media Bottles with HDPE Caps
SCI_ED

Catalog No. S139082 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S139082 1 L
S139079 125 mL
S139080 250 mL
S139081 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S139082 Supplier Eisco Labs Supplier No. CH5224PK12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile PET media bottles with secure HDPE caps, leakproof seals, and tamper-evident bands built for safe, reliable storage of lab reagents and buffers with lightweight durability and chemical resistance.

  • Sterile Media Bottles: PET bottles with secure HDPE caps, ideal for laboratory reagents and buffer storage.
  • Leakproof Design: Features semi-buttress threads and molded seal rings for a dependable, spill-free seal during storage and transport.
  • Tamper-evident Safety: Heat-shrink bands around each bottle ensure material security and protection from unauthorized access.
  • Durable and Lightweight: Made from robust PET with smooth, rounded edges for easy sample recovery and safe handling.
  • Wide Temperature Range: Suitable for storage between -70°C and +60°C, supporting diverse lab and biopharmaceutical applications.

Specifications

Capacity (Metric) 1 L
Color Clear
Diameter (Metric) Inner Neck 28.5 mm
Material PET
No. per Pack 12 Pk.
Sterility Sterile
Neck Finish 92.2 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 217.9 mm
Autoclavable Not Autoclavable
Capacity (English) 33.8 oz.
Shape Cylindrical
Disposable Single-use
Product Type Media Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.