Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco Labs Sterile PETG Media Bottles with HDPE Caps
SCI_ED

Catalog No. S139078 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
2 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S139078 2 L
S139074 30 mL
S139057 60 mL
S139073 125 mL
S139075 250 mL
S139076 500 mL
S139077 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. S139078 Supplier Eisco Labs Supplier No. CH5207PK6
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gamma-sterilized PETG media bottles feature secure HDPE caps, tamper-evident seals, and leak-proof threading are ideal for safely storing reagents and buffers in lab or biopharma settings with excellent durability and chemical resistance.

  • Sterile media bottles - PETG media bottles with secure HDPE caps. Ideal for storing lab reagents, buffers, and media in scientific or biopharmaceutical environments.
  • Leak proof and secure - Designed with semi-buttress threads and molded seal rings, ensuring a leak-proof closure for safe transport and storage of sensitive materials.
  • Gamma sterilized - Bottles are gamma sterilized per ISO 11137 standards, ensuring safe use for critical laboratory applications.
  • Durable and impact-resistant - PETG construction offers exceptional impact resistance, chemical compatibility, and thermal stability from -70°C to 60°C.
  • Tamper-evident safety - Each bottle features a tamper-evident heat-shrink seal for added security, ensuring product integrity during storage and use.

Specifications

Capacity (Metric) 2 L
Color Clear
Diameter (Metric) Inner Neck 39.75 mm
Material PETG
No. per Pack 6 Pk.
Sterility Sterile
Neck Finish 116.7 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 271.5 mm
Autoclavable Not Autoclavable
Capacity (English) 67.62 oz
Shape Cylindrical
Disposable Single-use
Product Type Media Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.