Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Sterile Phosphate Tuberculosis Buffer Solution
SDP

Catalog No. 22130511
Change view
Click to view available options
Sterile Phosphate Tuberculosis Buffer Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-130-511 Sterile Phosphate Tuberculosis Buffer Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-130-511 Supplier StatLab Supplier No. 669500
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Phosphate Buffer can be used for washing sputum after digestion and decontamination

  • Ready-to-use single bottle of sterilized buffer solution
  • No weighing, mixing, or autoclave time is needed for this ready-to-use sterilized buffer
  • Compatible with multiple automated TB systems
  • 500 mL bottle

Specifications

Chemical Name or Material Phosphate Buffers
For Use With (Application) Sputum Testing
Physical Form Solution
Quantity 500 mL
Formulation Liquid
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.