Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Sterile Pipette Tips
GREENER_CHOICESDP

Catalog No. 09035036 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
200 μL
300 μL
1000 μL
1200 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09-035-036 300 μL
09-035-032 1200 μL
09-035-033 1000 μL
09-035-037 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 09-035-036 Supplier VistaLab™ Supplier No. 40802033
Add to Cart
Add to Cart

Designed to provide a low-force tip solution for a more comfortable pipetting experience; less force needed to acquire tips

  • Designed to provide a low-force tip solution for a more comfortable pipetting experience. Less force needed to acquire tips.
  • Pipette tips are packaged in racks with hinged lids designed for easy one-handed opening.
  • 10 μL extended length tips have a maximum volume of 20 μL and are compatible with 2, 10, and 20 μL pipettes.
  • Fine tip for improved accuracy and precision.
  • Filter tips are fitted with a polymer barrier to reduce the risk of contamination and prevent aerosols from penetrating into the pipette.
  • Lids feature stacking supports, and are removable to save bench space.

Specifications

Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Format Hinged Rack Sterile
Tip Style Standard
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.