Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Sterile Scalpel Blades
SDP

Catalog No. 10001834 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
Size 10
Size 11
Size 12
Size 13
Size 14
Size 15
Size 15C
Size 18
Size 19
Size 21
Size 22
Size 23
Size 25A
For Use With (Equipment):
No. 3 and No. 7 handles
No. 4 handle
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Equipment)
10-001-834 Size 23 No. 4 handle
10-001-823 Size 11 No. 3 and No. 7 handles
10-001-824 Size 12 No. 3 and No. 7 handles
10-001-825 Size 13 No. 3 and No. 7 handles
10-001-826 Size 14 No. 3 and No. 7 handles
10-001-828 Size 15C No. 3 and No. 7 handles
10-001-829 Size 18 No. 4 handle
10-001-830 Size 19 No. 4 handle
10-001-833 Size 22 No. 4 handle
10-001-836 Size 25A No. 4 handle
10-001-983 Size 10 No. 3 and No. 7 handles
10-001-984 Size 15 No. 3 and No. 7 handles
10-001-986 Size 21 No. 4 handle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 10-001-834 Supplier Stoelting™ Supplier No. 5217523
Only null left
Add to Cart
Add to Cart

A variety of scalpel blades for surgical research needs

Specifications

For Use With (Application) Neuroscience
For Use With (Equipment) No. 4 handle
Size Size 23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.