Learn More
R&D Systems Sterile Single Cell Cartridge, Pack size: 10 cartridges, Dispensing volume: 1 μL, Maximum sample volume: 600 μL, Compatible with: 96-well and 384-well plates
SDP
Catalog No. NC2227836 Shop All Novus Products
Encompass
The ProteinSimple Sterile Single Cell Cartridge is designed for efficient and precise isolation of single cells needed for advanced genomics and cell biology applications. This cartridge guarantees sterile conditions and high reproducibility in single-cell experiments while being compatible with ProteinSimple Hana and Pala cell sorters. Key features include:
  • Precision single-cell isolation for multi-well plates
  • Compatible with various cell sorters and dispensers
  • Individually wrapped for sterility
  • Dispenses 1 cell per well
  • Maximum sample volume of 600 μL
  • Optimal dispensing rate of 1-2 events per second
  • Wide optimal cell concentration range of 5,000-20,000 cells/mL
  • Supports multiple plate types including 96-well and 384-well plates
  • High maximum cell concentration in enrichment workflows up to 150M cells/mL
This item is not returnable. View return policy
Catalog No. NC2227836 Supplier R&D Systems Supplier No. NC003
Customers require a valid business account associated to their profile to access pricing and ordering options.
The ProteinSimple Sterile Single Cell Cartridge is designed for efficient and precise isolation of single cells needed for advanced genomics and cell biology applications. This cartridge guarantees sterile conditions and high reproducibility in single-cell experiments while being compatible with ProteinSimple Hana and Pala cell sorters. Key features include:
  • Precision single-cell isolation for multi-well plates
  • Compatible with various cell sorters and dispensers
  • Individually wrapped for sterility
  • Dispenses 1 cell per well
  • Maximum sample volume of 600 μL
  • Optimal dispensing rate of 1-2 events per second
  • Wide optimal cell concentration range of 5,000-20,000 cells/mL
  • Supports multiple plate types including 96-well and 384-well plates
  • High maximum cell concentration in enrichment workflows up to 150M cells/mL