Learn More
Eisco Labs Sterile Square HDPE Reagent Bottles with Wide Mouth Polypropylene Caps
Shop All Eisco ProductsDescription
- Sterile reagent bottles - Sterile square HDPE bottles with polypropylene caps, designed for secure and versatile use in labs and industrial settings.
- Leak-proof design - Rigorously tested to prevent leaks under pressure, ensuring safe storage and transport of liquids and reagents.
- Excellent chemical resistance - Crafted from HDPE, offering high durability and compatibility with a variety of chemicals and reagents.
- Versatile lab and industrial use - Suitable for chemical storage, liquid handling, and mixing applications in research, laboratory, and industrial environments.
- Translucent and portable - Lightweight, translucent body for easy content visibility and a square design for optimal storage efficiency.
- Designed for chemical storage and transport, these leak-proof, chemical-resistant bottles are ideal for laboratory and industrial applications, -100°C to 120°C.
Specifications
Specifications
| Capacity (Metric) | 175 mL |
| Capacity (English) | 6 oz. |
| Color | Translucent |
| Material | HDPE |
| Sterility | Sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Product Type | Reagent Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.