Learn More
Eisco Labs Sterile Vented Polycarbonate Erlenmeyer Flasks with Baffled Bottom
Catalog No. 12141653 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-141-653 125 mL
12-141-633 250 mL
12-141-635 500 mL
12-141-654 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-141-653 Supplier Eisco Labs Supplier No. CH5281PK24
Only null left
Add to Cart
Add to Cart
Gamma sterilized and made from USP Class VI lab-grade polycarbonate for dependable laboratory performance.
  • Polycarbonate erlenmeyer flasks - Sterile polycarbonate flasks with vented caps for optimal gas exchange and controlled mixing.
  • Baffled bottom for enhanced mixing - Designed for efficient agitation, ideal for cell culture, fermentation, and suspension of particles.
  • Durable polycarbonate design - Highly impact-resistant and shatterproof, providing long-term reliability in demanding lab environments.
  • Gamma sterilized and lab compliant - Gamma sterilized per ISO 11137, USP Class VI compliant, and FDA 21 CFR 177.1630 certified for lab use.
  • Wide lab applications - Perfect for research, media preparation, biopharmaceutical processing, and use in orbital shakers.
Specifications
Capacity (Metric) 125 mL
Material Polycarbonate
Product Type Erlenmeyer Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Sterile
Bottom Shape Baffled