Learn More
Eisco Labs Sterile Vented Polycarbonate Erlenmeyer Flasks with Flat Bottom
Catalog No. 12141652 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
2000 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-141-652 2000 mL
12-141-649 125 mL
12-141-650 250 mL
12-141-631 500 mL
12-141-651 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-141-652 Supplier Eisco Labs Supplier No. CH5256PK04
Add to Cart
Add to Cart
Gamma sterilized and made from USP Class VI lab-grade polycarbonate for reliable laboratory use.
  • Polycarbonate Erlenmeyer Flasks - Sterile polycarbonate flasks with vented caps for gas exchange and controlled pressure.
  • Durable and Impact resistant - Polycarbonate construction offers high impact resistance, minimizing breakage in demanding lab environments.
  • Vented Cap for Gas Exchange - Allows airflow to prevent pressure buildup, ideal for biological cultures, fermentation, and media preparation.
  • Gamma Sterilized and Lab complaint - USP Class VI certified, ISO 11137 sterilized, FDA 21 CFR 177.1630 food contact compliant, and non-pyrogenic.
  • Versatile Lab applications - Designed for biopharmaceutical processing, cell culture, aeration-dependent applications, and chemical storage.
Specifications
Capacity (Metric) 2000 mL
Material Polycarbonate
Product Type Erlenmeyer Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Sterile
Bottom Shape Flat