Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Sterility Bottles

Catalog No. 22227315 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 mL
200 mL
300 mL
500 mL
Color:
Amber
Beige
Clear
Light Beige
Light Tan
Light Yellow
Media Type:
Fluid A
Fluid D
Fluid Thioglycollate Medium
Plate Count Agar
Potato Dextrose Agar
R2A Agar
Trypticase Soy Broth
Trypticase™ Soy Broth
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Media Type
22-227-315 500 mL Beige R2A Agar
DF9066-78-0 300 mL Clear Fluid D
DF9065-78-0 300 mL Light Yellow Fluid A
B299906 500 mL Light Tan Potato Dextrose Agar
B99107 100 mL Beige Trypticase Soy Broth
BB99417 100 mL Light Beige Fluid Thioglycollate Medium
L99112 500 mL Light Beige Fluid Thioglycollate Medium
B99108 100 mL Amber Fluid Thioglycollate Medium
DF9082-74 100 mL Clear Fluid A
B99113 500 mL Light Beige Trypticase™ Soy Broth
BB99094 200 mL Beige Plate Count Agar
DF9083-74 100 mL Clear Fluid D
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 22-227-315 Supplier BD Supplier No. 299436
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Certificate of Analysis

Glass sterility bottles available with a variety of media

  • Transparent labels for easy viewing and identification of medium
  • Labels are laminated acetate to resist alcohol or chlorine treatment for cleanroom use
  • Prepared bottles for sterility testing and microbial limits

Wide-Mouth Bottles

  • Openings are 48mm in diameter to accommodate bulky samples

Order Info

Supplied in packs of 10 bottles

Compliance

Conform to harmonized USP requirements.

TRUSTED_SUSTAINABILITY

Specifications

Cap Type Screw
Certifications/Compliance USP
Color Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type R2A Agar
Fill Volume 500 mL
Formulation Yeast extract 0.5g/L, Proteose Peptone No. 3 0.5g/L, Casamino Acids 0.5g/L, Dextrose 0.5g/L, Soluble Starch 0.5g/L, Sodium Pyruvate 0.3g/L, Dipotassium Phosphate 0.3g/L, Magnesium Sulfate 0.05g/L, Agar 15g/L
For Use With (Application) Enumerating heterotrophic organisms in treated potable Water
Includes Certificate of Analysis
Packaging Type Bottle
Quantity 500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.