Learn More
World Precision Instrument Sterilization Mesh Casette, 10.5x7x2.5cm
SDP
Supplier:  World Precision Instrument 504768
Encompass
Mesh sterilization baskets are perfect for sterilizing, storing and organizing small components or instrument. These baskets are manufactured from 304 stainlesss steel. They are non-magnetic and electropolished. They are burr-free to protect the instrument wraps from tearing. The fine mesh has open squares of 1.6 mm, and the wire edge diameter is 0.7 mm. These baskets come with a wire locking clip. This style of sterilization baskets is available in three sizes: 4x4x2 cm, 8x8x3.4 cm, and 10.5x7x2.5 cm.
Catalog No. 50-253-9290