Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS Coverage™ Spray TB Plus Ready-to-Use Disinfectant Cleaner

Catalog No. 1441513 Shop All STERIS Corporation Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
14-415-13 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-415-13 Supplier STERIS Supplier No. 162908

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Hard surface quaternary-based disinfectant kills a broad range of bacteria, viruses, and fungi within three minutes

Ideal for use in laboratories, hospitals, surgical centers, medical and dental offices, and other patient-care facilities.

  • Tuberculocidal and HIV-1 effective
  • Leaves no hazardous residue
  • Contains no dangerous phenols or chlorine
  • Compatible for use on inanimate surfaces including plastics, stainless steel, chrome, vinyl, acrylic, glass, Plexiglas™ resin, and baked-on painted surfaces

  • Uses a sprayer head on the 24 oz. (710mL) bottle that allows application without producing aerosolized airborne particles that may be aspirated

Compliance

Conforms to OSHA Guidelines (21CFR Part 1910, 1030).

Specifications

Form Liquid
pH 6.5 to 8.7
Appearance Clear
Certifications/Compliance OSHA
Composition 21% Isopropanol
Container Type Refill bottle
For Use With (Application) Kills a broad range of Bacteria, Viruses and Fungi on inanimate surfaces including Plastics, Stainless Steel, Chrome, Vinyl, Acrylic, Glass, Plexiglas and baked-on painted surfaces
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.