Learn More
STERIS Process Vesphene™ II st Sterile One-step Cleaner Disinfectant
Shop All STERIS Corporation ProductsSpecifications
Specifications
| Product Type | Cleaner/Disinfectant |
| pH | 10.4 to 10.6 |
| Sterility | Sterile |
| Container Type | Pouch |
| Packaging | Double bagged |
| Volume (English) | 2 oz. |