Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Steritop™ Threaded Bottle Top Filters

Catalog No. S2GPT10RE Shop All MilliporeSigma Products
Change view
Click to view available options
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
Material (Membrane):
PES
PVDF
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Material (Membrane)
S2GPT10RE 1000 mL PES
S2GPT01RE 150 mL PES
S2GPT02RE 250 mL PES
S2GPT05RE 500 mL PES
S2GVT05RE 500 mL PVDF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. S2GPT10RE Supplier MilliporeSigma™ Supplier No. S2GPT10RE
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.

  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.

Order Info

Note: Image above shows Bottle Top Filter with receiver flask (sold separately).

TRUSTED_SUSTAINABILITY

Specifications

Color Clear with blue adapter
Disposable Single-use
Material Polystyrene
Material (Membrane) PES
Pore Size 0.22 μm
Sterility Sterile
For Use With (Application) Sterile filtration
Outlet Fitting 45 mm
Type Bottle Top Filter
Volume (Metric) 1000 mL
Length (Metric) 169 mm
Height (Metric) 169 mm
Diameter (Metric) 45 mm
Diameter (Metric) Filter 13.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.