Learn More
Description
- Ideal for the classroom, especially in the study of respiration and circulation
- Teaching stethoscope has 3.8 cm Bowles chestpiece and two sets of binaurals joined together so the teacher and student can listen simultaneously
- Bowles-type stethoscope has adjustable, chrome-plated binaurals and PVC Y tubing
Specifications
Specifications
| Product Type | Dual Earpiece-Teaching Stethoscope |
| Color | Black |
| For Use With (Application) | For the study of respiration and circulation in the classroom |
| Includes | 3.8 cm Bowles Chestpiece, Two Sets of Adjustable Chrome Plated Binaurals, PVC Y-tubing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

