Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Stevens Scissors
SDP

Catalog No. 22445541 Shop All Mopec Products
Change view
Click to view available options
Length (English):
3.75 in.
4.125 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-445-541 4.125 in.
22-445-547 3.75 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-445-541 Supplier Mopec™ Supplier No. AA436
Add to Cart
Add to Cart

Used for cutting with extreme precision in small incisions.

  • All micro scissors are made from stainless steel

Specifications

Blade Material Stainless Steel
Product Type Micro Scissors
Blade Style Straight
Color Code Gray
Color-coded No
Grade Standard
Handle Material Stainless Steel
Length (English) 4.125 in.
Length (Metric) 10.48 cm
Material Stainless Steel
Plug Type US-style
Size Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.