Learn More
Selleck Chemical LLC STF-62247 10mg 315702-99-9
SDP
Supplier:  Selleck Chemical LLC S104110MG
Encompass_Preferred
STF-62247 is a molecule targeting VHL-deficient renal cell carcinoma that induces autophagy. STF-62247 shows selective toxicity and growth inhibition of renal cells lacking VHL; 25-fold greater sensitivity observed for cells with VHL deficiency compared to wild-type (VHL+). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-759-1
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.