Learn More
New Pig Sticky Steps™ Mat with Frame
SDP
Catalog No. 18389136 Shop All New Pig Corporation Products
Change view
Click to view available options
Color:
White
Catalog No. Color
18-389-136 White
1 options
This item is not returnable. View return policy
Catalog No. 18-389-136 Supplier New Pig Supplier No. MAT56824X36ENG
Add to Cart
Add to Cart
This item is not returnable. View return policy
This tacky-surface floor mat in a tray is portable, so you can move it anywhere you need it.
  • High-tack disposable mat pulls dirt and debris from shoes and wheels to prevent tracking into offices
  • Mats come 30 sheets to a pad; entire pad attaches to frame and used sheets tear off easily
  • Numbered pull-tab system shows when you're running low
  • Each 4 mil polyethylene sheet is nonallergenic, nonodorous and nondrying
  • Mats on frame are clear (refills sold separately, see MAT195)
  • Refillable white PVC frame with rubber backing holds the mat pad in place Portable frame lets you use
  • Sticky Steps Mat on carpet or on floors that are difficult to clean thoroughly; can be easily stored Ideal for use in hospitals, laboratories, clean rooms, data centers, test facilities, mixing facilities, construction sites, residences, photography studios and during renovation projects
Specifications
Color White
Material Polyethylene