Learn More
New Pig Sticky Steps™ Mat
SDP
Catalog No. 18389131 Shop All New Pig Corporation Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
18-389-131 Gray
1 options
This item is not returnable. View return policy
Catalog No. 18-389-131 Supplier New Pig Supplier No. MAT195GY
Add to Cart
Add to Cart
This item is not returnable. View return policy
Multi-sheet floor mat grabs grime from shoes and wheels to keep dirt out of offices, cleanrooms and other areas where clean floors are vital.
  • High-tack disposable mat pulls dirt and debris from shoes and wheels to prevent tracking into offices
  • Used sheets tear off easily and individuall
  • Numbered pull-tab system shows when you're running low
  • Low-profile pack adheres to floor with easy-to-use backing
  • Each 4 mil polyethylene sheet is nonallergenic, nonodorous and nondrying
Specifications
Color Gray
Material Polyethylene