Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Sting Relief Pads

Catalog No. 19090686 Shop All Medique Products
Catalog No. 19-090-686 Supplier Medique Supplier No. 23112
Add to Cart
Add to Cart

For relief of pain from insect bites and stings, minor scrapes and minor pains.

  • Instant cold compress

Specifications

For Use With (Application) Relief of pain from insect bites and stings, minor scrapes and minor pains.
Disposable Single-use
Format Pad
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.