Learn More
United Scientific™ Stir Bar Assortment
SDPSCI_ED
Catalog No. S99478 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
1 Set of 20 Bars
1 Set of 50 Bars
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S99478 1 Set of 20 Bars
S99479 1 Set of 50 Bars
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S99478 Supplier United Scientific™ Supplier No. MSBA20
Only null left
Add to Cart
Add to Cart
Stir Bar Assortment made from high-quality magnets coated with PTFE are for use with hot plates/stirrers.
Available in sets of 20 and sets of 50.
Set of 20 includes:
  • 2 octagon-shaped stir bars with pivot ring, 15 mm x 7.5 mm
  • 2 octagon-shaped stir bars with pivot ring, 25 mm x 8 mm
  • 2 octagon-shaped stir bars with pivot ring, 35 mm x 9 mm
  • 2 octagon-shaped stir bars with pivot ring, 40 mm x 8 mm
  • 2 octagon-shaped stir bars with pivot ring, 48.5 mm x 10 mm
  • 2 polygon-shaped stir bars without pivot ring, 12 mm x 3 mm
  • 2 polygon-shaped stir bars without pivot ring, 70 mm x 9 mm
  • 2 egg-shaped stir bars with blunt edges, 15 mm x 7 mm
  • 2 egg-shaped stir bars with blunt edges, 30 mm x 13 mm
  • 2 egg-shaped stir bars with blunt edges, 50 mm x 20 mm
Set of 50 includes:
  • 5 octagon-shaped stir bars with pivot ring, 15 mm x 7.5 mm
  • 5 octagon-shaped stir bars with pivot ring, 25 mm x 8 mm
  • 5 octagon-shaped stir bars with pivot ring, 35 mm x 9 mm
  • 5 octagon-shaped stir bars with pivot ring, 40 mm x 8 mm
  • 5 octagon-shaped stir bars with pivot ring, 48.5 mm x 10 mm
  • 5 polygon-shaped stir bars without pivot ring, 12 mm x 3 mm
  • 5 polygon-shaped stir bars without pivot ring, 70 mm x 9 mm
  • 5 egg-shaped stir bars with blunt edges, 15 mm x 7 mm
  • 5 egg-shaped stir bars with blunt edges, 30 mm x 13 mm
  • 5 egg-shaped stir bars with blunt edges, 50 mm x 20 mm
Specifications
Material PTFE
Length (Metric) Various
Shape Various
Dimensions (L x W x H) Various
Quantity 20 Pk.
Type Stir Bar
Coating PTFE
Includes 1 Set of 20 Bars
Autoclavable Autoclavable
Color White