Learn More
V&P Scientific Inc Stirrer Accessory Kit, includes Stand, Arm, Clamp and Plastic Syringe Holder
Supplier:  V&P Scientific Inc VP 710D34
Encompass
STIRRER ACCESSORY, Kit for Syringe Pumps, includes Lab Support Stand, Aluminum Arm (Attaches to VP 710D3), Clamp, and Plastic Syringe Holder for KD Scientific Pump(Legato210), plus 2 Phillips Head Screws 8-32 x 5/8"
Catalog No. NC2395335
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.