Learn More
DWK Life Sciences Kimble™ Stirrer Assembly with Stainless-Steel Blade and Shaft
SDP
Catalog No. K7850400129 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
Joint: 29/42
Joint: 45/50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
K7850400129 Joint: 29/42
K7850400145 Joint: 45/50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K7850400129 Supplier DWK Life Sciences Supplier No. 7850400129
Complete unit includes a Hi-Vac™ PTFE and glass stirrer bearing and 3/8 in. (10mm) O.D. stainless-steel stir shaft/blade for high torque applications. This assembly is recommended for systems requiring vacuum.
TRUSTED_SUSTAINABILITY
Specifications
Standard Taper Joint Joint: 29/42