Learn More
Hach Company Stirrer, Mixer, Hand-Held
Supplier: Hach Company 2496400
Hand-held mini-mixer used with test tubes, micro centrifuge tubes and containers too narrow for conventional mixers. Stirring rod flares out for use in larger spaces.Clear vinyl carrying case, 2 AA batteries included.Stirring rod dimension is 11.4 x 6.4cm (4.5" x 0.25 inches)Dimensions including rod 20.3 x 3.2 x 2.5 cmWeight 3 oz.